DETA NONOate (NOC-18) 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
DETA/NO, 2,2′-(Hydroxynitrosohydrazino)bis-ethanamine, Diethylenetriamine NONOate, 3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene
32865-0250
32865-1000
TS32865-0250EA
99
USD
TS32865-0250
TS32865-1000
DETA NONOate (NOC-18) 95%
DETA NONOate (NOC-18)
Formula:
C₄H₁₃N₅O₂ Melting Pt: 98…106 °C Storage Temperature: Freezer |
MDL Number:
MFCD00278808 CAS Number: 146724-94-9 |
Specification Test Results
Appearance | White to off-white Crystalline solid |
Infrared spectrum | Conforms |
NMR | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...