Indole-3-carboxylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Indole-3-carboxylic acid

30761-0010 30761-0050
TS30761-0010EA 40 USD
TS30761-0010 TS30761-0050
Indole-3-carboxylic acid 99%
Indole-3-carboxylic acid
Formula: C₉H₇NO₂
Melting Pt: 200…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00005624
CAS Number: 771-50-6
EINECS: 212-231-6

Order Now

Specification Test Results

Appearance Light beige Powder
Infrared spectrum Conforms
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR