(S)-(+)-Camptothecin 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione

27672-0010 27672-1000
TS27672-0010EA 696 USD
TS27672-0010 TS27672-1000
(S)-(+)-Camptothecin 98%
(S)-(+)-Camptothecin
Formula: C₂₀H₁₆N₂O₄
MW: 348.36 g/mol
Melting Pt: 260 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00081076
CAS Number: 7689-03-4
UN: 1544
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Loss on drying ≤1.0 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR