D-Glucono-1,5-lactone 99%

Supplier: Thermo Scientific Chemicals

27105-0010 27105-0051 27105-2500
TS27105-0010EA 87 USD
TS27105-0010 TS27105-0051 TS27105-2500
D-Glucono-1,5-lactone 99%
D-Glucono-1,5-lactone
Formula: C₆H₁₀O₆
Melting Pt: 151…155 °C (dec.)
Density: 0.6 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00006647
CAS Number: 90-80-2
EINECS: 202-016-5
Merck Index: 13,04470

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 151 to 158 °C (Dec)
Ash ≤0.1 %
Titration excess NaOH ≥98.5 %
Lead (Pb) ≤10 ppm
Heavy metals (as Pb) ≤20 ppm
Reducing substances ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR