Ethyl-3-(dimethylamino)acrylate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 3-(dimethylamino)acrylate, 3-(Dimethylamino)acrylic Acid Ethyl Ester, Ethyl 3-(N,N-dimethylamino)acrylate

26963-0250 26963-1000
TS26963-0250EA 80 USD
TS26963-0250 TS26963-1000
Ethyl-3-(dimethylamino)acrylate 99+%
Ethyl-3-(dimethylamino)acrylate
Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
Boiling Pt: 118…121 °C (10HPA)
Melting Pt: 17…18 °C
Density: 1 g/cm³
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00144269
CAS Number: 924-99-2

Order Now

Specification Test Results

Appearance Clear yellow to light orange Liquid
Infrared spectrum Conforms
Refractive index 1.5105 to 1.5125 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR