Theobromine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,7-Dimethyl-3,7-dihydro-1H-purine-2,6-dione, 3,7-Dimethylxanthine, 3,7-Dihydro-3,7-dimethyl-1H-purine-2,6-dione, 2,6-Dihydroxy-3,7-dimethylpurine

25882-0010 25882-1000 25882-2500
TS25882-0010EA 501 USD
TS25882-0010 TS25882-1000 TS25882-2500
Theobromine 99%
Theobromine
Formula: C₇H₈N₄O₂
MW: 180.17 g/mol
Melting Pt: 290…295 °C
Storage Temperature: Ambient
MDL Number: MFCD00022830
CAS Number: 83-67-0
EINECS: 201-494-2
Merck Index: 12,09418

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C)
Titration with NaOH ≥98.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR