L-Dehydroascorbic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(5R)-5-[(1S)-1,2-dihydroxyethyl]oxolane-2,3,4-trione, L-Dehydroascorbate, L-Threo-hexo-2,3-diulosono-1,4-lactone
25093-0010
25093-0050
25093-2500
TS25093-0010EA
157
USD
TS25093-0010
TS25093-0050
TS25093-2500
L-Dehydroascorbic acid 96%
L-Dehydroascorbic acid
Formula:
C₆H₆O₆ MW: 174.108 g/mol Melting Pt: 228 °C Storage Temperature: Freezer |
MDL Number:
MFCD00066467 CAS Number: 490-83-5 |
Specification Test Results
Appearance | White to light brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥95.0 % (ELSD) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...