2-Hydroxynicotinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxypyridine-3-carboxylic acid, 1,2-Dihydro-2-oxonicotinic acid, 2-Hydroxy-3-pyridinecarboxylic acid, 2-Oxo-1,2-dihydropyridine-3-carboxylic acid

24722-0250 24722-1000
TS24722-0250EA 51 USD
TS24722-0250 TS24722-1000
2-Hydroxynicotinic acid 98%
2-Hydroxynicotinic acid
Formula: C₆H₅NO₃
Melting Pt: 259…261 °C
MDL Number: MFCD00010100
CAS Number: 609-71-2
EINECS: 210-198-2

Order Now

Specification Test Results

Appearance White to cream to light yellow Powder
Infrared spectrum Conforms
Melting point 256 to 262 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR