Potassium bromide 99+% for biochemistry
Supplier: Thermo Scientific Chemicals
|
Warning
|
21258-0010
21258-2500
TS21258-0010EA
279
USD
TS21258-0010
TS21258-2500
Potassium bromide 99+% for biochemistry
Potassium bromide
Formula:
KBr Boiling Pt: 1435 °C (760 mmHg) Melting Pt: 734 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011358 CAS Number: 7758-02-3 |
Specification Test Results
Appearance | White to off-white Crystalline powder or crystals |
Titration Argentometric | ≥99.0 % |
Heavy metals | ≤5 ppm |
Chloride (Cl) | ≤0.2 % |
Iodide (I) | ≤0.001 % |
Bromate (BrO3) | ≤0.001 % |
Iodate (IO3) | ≤0.001 % |
Sulfate (SO4) | ≤0.005 % |
Barium (Ba) | ≤0.002 % |
Calcium (Ca) | ≤0.002 % |
Iron (Fe) | ≤0.0005 % |
Magnesium (Mg) | ≤0.001 % |
Sodium (Na) | ≤0.02 % |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...