Zein, purified

Supplier: Thermo Scientific Chemicals

17931-5000 17931-0025 17931-1000
TS17931-5000EA 132 USD
TS17931-5000 TS17931-0025 TS17931-1000
Zein, purified
Zein
Storage Temperature: Ambient
MDL Number: MFCD00062414
CAS Number: 9010-66-6

Order Now

Specification Test Results

Appearance Yellow to straw yellow to tan Powder
Infrared spectrum Conforms
Loss on drying ≤8.0 % (105°C, 2 h)
Heavy metals ≤20 ppm
Protein 88.0 to 96.0 % (On dried basis)
Residue after ignition ≤2.0 % (800°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR