Quercetin hydrate 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,3',4',5,7-Pentahydroxyflavone hydrate, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one xhydrate

17407-0100 17407-0250 17407-1000
TS17407-0100EA 33 USD
TS17407-0100 TS17407-0250 TS17407-1000
Quercetin hydrate 95%
Quercetin hydrate

Appearance: Yellow Crystalline powder

Formula: C₁₅H₁₀O₇
MW: 302.24 g/mol (anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006828
CAS Number: 849061-97-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Water ≤12 % (K.F.)
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR