Succinonitrile 99+%

Supplier: Thermo Scientific Chemicals
Warning

17118-1000 17118-5000
TS17118-1000EA 46 USD
TS17118-1000 TS17118-5000
Succinonitrile 99+%
Succinonitrile
Formula: C₄H₄N₂
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 57 °C
Flash Pt: 132 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001949
CAS Number: 110-61-2
EINECS: 203-783-9
Merck Index: 12,09041

Order Now

Specification Test Results

Appearance Colorless to white to light yellow Crystalline mass or waxy solid
Infrared spectrum Conforms
GC ≥96 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR