Isophthalonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Benzenedicarbonitrile, 1,3-Dicyanobenzene, Benzene-1,3-dicarbonitrile

16366-0050 16366-2500
TS16366-0050EA 27 USD
TS16366-0050 TS16366-2500
Isophthalonitrile 98%
Isophthalonitrile
Formula: C₈H₄N₂
Boiling Pt: 288 °C (1013 hPa)
Melting Pt: 161…165 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00001795
CAS Number: 626-17-5
EINECS: 210-933-7
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to tan Flakes
Infrared spectrum Conforms
Melting point 159 to 163 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR