4-(Indol-3-yl)butyric acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Indolebutyric acid, Indole-3-butyric acid

16069-0050 16069-0250 16069-1000
TS16069-0050EA 47 USD
TS16069-0050 TS16069-0250 TS16069-1000
4-(Indol-3-yl)butyric acid 98%
4-(Indol-3-yl)butyric acid
Formula: C₁₂H₁₃NO₂
Melting Pt: 120…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005664
CAS Number: 133-32-4
EINECS: 205-101-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,04987

Order Now

Specification Test Results

Appearance White to yellow to beige to brown Powder
Infrared spectrum Conforms
Melting point 120 to 125 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR