N-Methyl-1,3-propanediamine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Methylpropane-1,3-diamine, 3-Methylamino propylamine

15557-0010 15557-0025 15557-2500
TS15557-0010EA 198 USD
TS15557-0010 TS15557-0025 TS15557-2500
N-Methyl-1,3-propanediamine 99%
N-Methyl-1,3-propanediamine
Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Boiling Pt: 139…141 °C (760 mmHg)
Melting Pt: –72 °C
Density: 0.84 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00008209
CAS Number: 6291-84-5
EINECS: 228-544-6
UN: 2922
ADR: 8 I

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4458 to 1.4478 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.5 %
Color scale ≤50 APHA
Titration with HCl ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR