N-Methylethylenediamine 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Aminoethylmethylamine, N1-Methylethane-1,2-diamine

15556-0050 15556-0250 15556-1000
TS15556-0050EA 74 USD
TS15556-0050 TS15556-0250 TS15556-1000
N-Methylethylenediamine 95%
N-Methylethylenediamine
Formula: C₃H₁₀N₂
MW: 74.13 g/mol
Boiling Pt: 114
Density: 0.84 g/cm³
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00008165
CAS Number: 109-81-9
EINECS: 203-707-4
UN: 2734
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4390 to 1.4420 (20°C, 589 nm)
Titration with HClO4 ≥94.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR