Acetamide 99%, pure

Supplier: Thermo Scientific Chemicals
Warning

15363-0025 15363-1000 15363-5000
TS15363-0025EA 255 USD
TS15363-0025 TS15363-1000 TS15363-5000
Acetamide 99%, pure
Acetamide
Formula: C₂H₅NO
Boiling Pt: 221 °C (760 mmHg)
Melting Pt: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00008023
CAS Number: 60-35-5
EINECS: 200-473-5
Merck Index: 13,00044

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
Melting point 78 to 81 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR