Amantadine hydrochloride 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Adamantylamine hydrochloride, 1-Aminotricyclo[3.3.1.1(3,7)]decane hydrochloride, 1-Aminotricyclo(3.3.1.1(3,7))decan HCl, 1-Aminoadamantane hydrochloride, 1-Adamantanammonium chloride, 1-Adamantanamine hydrochloride, Tricyclo[3.3.1.1(3,7)]dec-1-ylamine hydrochloride, Adamantan-1-amine hydrochloride, 1-Adamantaneammonium Chloride

15167-0250 15167-1000
TS15167-0250EA 73 USD
TS15167-0250 TS15167-1000
Amantadine hydrochloride 99+%
Amantadine hydrochloride
Formula: C₁₀H₁₈ClN
MW: 187.71 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00074723
CAS Number: 665-66-7
EINECS: 211-560-2

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥99.0 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR