Acetanilide 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Phenylacetamide

15081-0010 15081-0050 15081-0051 15081-2500 15081-5000
TS15081-0010EA 125 USD
TS15081-0010 TS15081-0050 TS15081-0051 TS15081-2500 TS15081-5000
Acetanilide 99+%
Acetanilide
Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 304 °C (760 mmHg)
Melting Pt: 111…115 °C
Flash Pt: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD00008674
CAS Number: 103-84-4
EINECS: 203-150-7
Merck Index: 13,00051

Order Now

Specification Test Results

Appearance White to beige to grey to light brown Powder or crystals or flakes
Infrared spectrum Conforms
Melting point 111 to 115 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR