Urea 99.5% for analysis

Supplier: Thermo Scientific Chemicals

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

14075-0010 14075-0050 14075-0250
TS14075-0010EA 75 USD
TS14075-0010 TS14075-0050 TS14075-0250
Urea 99.5% for analysis
Urea
Formula: NH₂CONH₂
MW: 60.06 g/mol
Boiling Pt: 197
Melting Pt: 132.7
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
Melting point 132 to 135 °C
Residue after ignition ≤0.01 %
Titration with HClO4 ≥99.4 %
Chloride (Cl) ≤5 ppm
Sulfate (SO4) ≤10 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR