Thiourea 99%, extra pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sulfourea, Thiocarbamide, Sulphourea

13891-0010 13891-0050 13891-5000
TS13891-0010EA 88 USD
TS13891-0010 TS13891-0050 TS13891-5000
Thiourea 99%, extra pure
Thiourea
Formula: NH₂CSNH₂
MW: 76.12 g/mol
Melting Pt: 171…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00008067
CAS Number: 62-56-6
EINECS: 200-543-5
UN: 3077
ADR: 9 III
Merck Index: 13,09443

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 171 to 177 °C
Titration Iodimetric ≥98.5 %
Loss on drying ≤0.5 %
Sulfated ash ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR