5-Aminosalicylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Mesalamine, Mesalazine, 5-Amino-2-hydroxybenzoic acid, m-Aminosalicylic acid

13433-0050 13433-0250 13433-1000
TS13433-0050EA 27 USD
TS13433-0050 TS13433-0250 TS13433-1000
5-Aminosalicylic acid 99%
5-Aminosalicylic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 283 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00007877
CAS Number: 89-57-6
EINECS: 201-919-1
Merck Index: 12,05964

Order Now

Specification Test Results

Appearance Beige to grey to light brown Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C, 3 hrs)
HPLC ≥98.5 %
Titration with HClO4 ≥98.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR