Rutin hydrate 97+%

Supplier: Thermo Scientific Chemicals

13239-0050 13239-1000 13239-5000
TS13239-0050EA 22 USD
TS13239-0050 TS13239-1000 TS13239-5000
Rutin hydrate 97+%
Rutin hydrate
Formula: C₂₇H₃₀O₁₆
Melting Pt: 195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006830
CAS Number: 207671-50-9

Order Now

Specification Test Results

Appearance Yellow to yellow-green to straw yellow Powder
Infrared spectrum Conforms
Loss on drying 5.0 % to 9.0 % (500 mg, 105°C, 3 h)
Ash ≤0.5 %
Titration with NaOH 97.0 to 102.0 % (On dry basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR