O-Phenylenediamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2-Phenylendiamine, 1,2-Phenylenediamine, 1,2-Diaminobenzene, OPD

13055-0010 13055-0050 13055-2500
TS13055-0010EA 140 USD
TS13055-0010 TS13055-0050 TS13055-2500
O-Phenylenediamine 98%
o-Phenylenediamine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 257
Melting Pt: 103
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00007721
CAS Number: 95-54-5
EINECS: 202-430-6
UN: 1673
ADR: 6.1 III
Merck Index: 12,07438

Order Now

Specification Test Results

Appearance Tan to brown Crystalline powder or crystals or flakes (May darken on storage)
Infrared spectrum Conforms
Melting point 100 to 104 °C
GC ≥97.5 %
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR