Pentaerythritol 98%

Supplier: Thermo Scientific Chemicals

12987-0010 12987-0050 12987-2500
TS12987-0010EA 44 USD
TS12987-0010 TS12987-0050 TS12987-2500
Pentaerythritol 98%
Pentaerythritol
Formula: C₅H₁₂O₄
MW: 136.15 g/mol
Boiling Pt: 380
Melting Pt: 257…264 °C
Flash Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00004692
CAS Number: 115-77-5
EINECS: 204-104-9
Merck Index: 12,07245

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
Melting point 257 to 264 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR