N-Methylaniline 99%

Supplier: Thermo Scientific Chemicals
Danger

12627-0010 12627-0025 12627-5000
TS12627-0010EA 73 USD
TS12627-0010 TS12627-0025 TS12627-5000
N-Methylaniline 99%
N-Methylaniline
Formula: C₇H₉N
Boiling Pt: 195 °C (760 mmHg)
Melting Pt: –57 °C
Density: 0.987 g/cm³
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00008283
CAS Number: 100-61-8
EINECS: 202-870-9
UN: 2294
ADR: 6.1,III
Merck Index: 12,06098

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5685 to 1.5725 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤20 APHA
Residue after evaporation ≤0.003 %
Carbonyl compounds ≤0.01 % (as formaldehyde)
Titrable acid ≤0.001 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR