(±)-Mandelic acid 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Hydroxy-2-phenylacetic acidDL-2-Hydroxy-2-phenylacetic acidDL-Amygdalic AcidDL-Mandelic acidMandelic acid(±)-2-Hydroxy-2-phenylacetic acid

12531-0050 12531-1000 12531-0010 12531-2500
TS12531-0050EA 465 USD
TS12531-0050 TS12531-2500 TS12531-1000 TS12531-0010
(±)-Mandelic acid 99+%
(±)-Mandelic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00064250
CAS Number: 90-64-2
EINECS: 210-277-1

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 117 to 121 °C
Titration with NaOH ≥99.0 %
Nitrate (NO3) ≤0.001 %
Phosphate (PO4) ≤0.0003 %
Aluminium (Al) ≤0.001 %
Arsenic (As) ≤0.00002 %
Cadmium (Cd) ≤0.0005 %
Calcium (Ca) ≤0.001 %
Chromium (Cr) ≤0.0005 %
Cobalt (Co) ≤0.0005 %
Copper (Cu) ≤0.0005 %
Iron (Fe) ≤0.002 %
Lead (Pb) ≤0.0005 %
Magnesium (Mg) ≤0.0005 %
Manganese (Mn) ≤0.0005 %
Nickel (Ni) ≤0.0005 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.005 %
Zinc (Zn) ≤0.0005 %
Heavy metals (as Pb) ≤20 ppm
Insoluble matter ≤0.005 % (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR