Malononitrile 99%

Supplier: Thermo Scientific Chemicals
Danger

12527-0025 12527-1000 12527-5000
TS12527-0025EA 328 USD
TS12527-0025 TS12527-1000 TS12527-5000
Malononitrile 99%
Malononitrile
Formula: C₃H₂N₂
Boiling Pt: 220 °C (760 mmHg)
Melting Pt: 30…34 °C
Density: 1.04 g/cm³
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00001883
CAS Number: 109-77-3
EINECS: 203-703-2
UN: 2647
ADR: 6.1,II
Merck Index: 13,05733

Order Now

Specification Test Results

Appearance White to yellow-brown Low melting crystalline mass
Infrared spectrum Conforms
Melting point 30 to 34 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR