Indole-5-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Indole-5-carboxylic acid

12221-0010 12221-0050
TS12221-0010EA 46 USD
TS12221-0010 TS12221-0050
Indole-5-carboxylic acid 98%
Indole-5-carboxylic acid
Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 207…212 °C
MDL Number: MFCD00005678
CAS Number: 1670-81-1
EINECS: 216-799-6

Order Now

Specification Test Results

Appearance Yellow to light beige Powder
Infrared spectrum Conforms
Melting point 207 to 212 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR