Indol-3-acetic acid 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: IAA, 1H-Indole-3-acetic acid, Heteroauxin, 3-Indoleacetic acid

12216-0100 12216-0250 12216-1000 12216-5000
TS12216-0100EA 44 USD
TS12216-0100 TS12216-0250 TS12216-1000 TS12216-5000
Indol-3-acetic acid 99+%
Indol-3-acetic acid
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 167 °C
Flash Pt: 171 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005636
CAS Number: 87-51-4
EINECS: 201-748-2
Merck Index: 13,04986

Order Now

Specification Test Results

Appearance White to beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 165 to 169 °C
HPLC ≥99.0 %
Elemental Analysis 3.72 to 3.99 % N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR