Hypoxanthine 99.5%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Purin-6-one, 1,9-Dihydro-6H-purin-6-one, Purin-6(1H)-one, 6-Purinone

12201-0250 12201-1000 12201-0050
TS12201-0250EA 98 USD
TS12201-0250 TS12201-1000 TS12201-0050
Hypoxanthine 99.5%
Hypoxanthine
Formula: C₅H₄N₄O
MW: 136.11 g/mol
Melting Pt: >250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005725
CAS Number: 68-94-0
EINECS: 200-697-3
Merck Index: 13,04895

Order Now

Specification Test Results

Appearance White to light yellow to light beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
Sulfated ash ≤0.2 %
HPLC ≥99.4 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR