Fumaric acid 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: trans-1,2-Ethylentricarboxylic acid

11975-0010 11975-0025 11975-0100 11975-1000
TS11975-0010EA 55 USD
TS11975-0010 TS11975-0025 TS11975-0100 TS11975-1000
Fumaric acid 99+%
Fumaric acid
Formula: C₄H₄O₄
MW: 116.07 g/mol
Boiling Pt: 356
Melting Pt: 287 (subl.)
Density: 1.62 g/cm³
Flash Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00002700
CAS Number: 110-17-8
EINECS: 203-743-0
Merck Index: 13,04308

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 295 to 300 °C
Water ≤0.25 % (K.F.)
HPLC ≥99.0 %
Titration with NaOH 99.0 to 101.0 % (On anhydrous substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR