Eugenol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole

11911-1000 11911-5000 11911-0025 11911-0050
TS11911-1000EA 49 USD
TS11911-1000 TS11911-5000 TS11911-0025 TS11911-0050
Eugenol 99%
Eugenol
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 255
Melting Pt: -10
Density: 1.06 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00008654
CAS Number: 97-53-0
EINECS: 202-589-1
Merck Index: 13,03931

Order Now

Specification Test Results

Appearance Clear colorless to yellow-green Liquid
Infrared spectrum Conforms
Refractive index 1.5400 to 1.5420 (20°C, 589 nm)
GC ≥98.5 %
Specific gravity (20°C) 1.064 to 1.070


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR