2-Ethylaniline 98%

Supplier: Thermo Scientific Chemicals
Danger

11805-0010 11805-0050 11805-2500
TS11805-0010EA 198 USD
TS11805-0010 TS11805-0050 TS11805-2500
2-Ethylaniline 98%
2-Ethylaniline
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 210
Melting Pt: -46
Density: 0.98 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007751
CAS Number: 578-54-1
EINECS: 209-424-2
UN: 2273
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Clear yellow to red-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5590 to 1.5610 (20°C, 589 nm)
GC ≥97.5 %
Chloride (Cl) ≤0.001 %
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤0.002 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤0.002 %
Potassium (K) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %
KMnO4-reducing substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR