Phosphonitrilic chloride trimer 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Cyclic Phosphonitrilic Chloride Trimer, Hexachlorocyclotriphosphazene

OR30304-100G OR30304-25G OR30304-500G
76616-322EA 80.48 USD
76616-322 76616-324 76616-326
Phosphonitrilic chloride trimer 97%
Phosphonitrilic chloride trimer
Formula: Cl₆N₃P₃
Boiling Pt: 127 °C/13mmHg
Melting Pt: 111…115 °C
Density: 1.98 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006474
CAS Number: 940-71-6
EINECS: 213-376-8
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR