N,N'-Bis(2-aminoethyl)-1,3-propanediamine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3,7-Diaza-1,9-nonanediamine, N1,N1'-(Propane-1,3-diyl)bis(ethane-1,2-diamine), 1,4,8,11-Tetraazaundecane

OR10977-100G OR10977-25G OR10977-5G
76610-982EA 192.54 USD
76610-982 76610-984 76610-986
N,N'-Bis(2-aminoethyl)-1,3-propanediamine
N,N'-Bis(2-aminoethyl)-1,3-propanediamine
Formula: C₇H₂₀N₄
Boiling Pt: 280 °C
Density: 0.96 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00008174
CAS Number: 4741-99-5
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR