6-Thioguanine

Supplier: APOLLO SCIENTIFIC
Danger

OR4016-1G OR4016-5G
76616-678EA 20.28 USD
76616-678 76616-680
6-Thioguanine
6-Thioguanine
Formula: C₅H₅N₅S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00233553
CAS Number: 154-42-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR