6-Chloro-7-deazapurine
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR2836-25G
OR2836-5G
76616-222EA
28.52
USD
76616-222
76616-224
6-Chloro-7-deazapurine
6-Chloro-7-deazapurine
Formula:
C₆H₄ClN₃ Melting Pt: 188…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD01686865 CAS Number: 3680-69-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...