2,6-Dichloropyridine-4-methanol 95+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,6-Dichloro-4-pyridinemethanol, 2,6-Dichloro-4-(hydroxymethyl)pyridine

OR0150-1G OR0150-5G
76615-148EA 209.09 USD
76615-148 76615-150 76615-152
2,6-Dichloropyridine-4-methanol 95+%
2,6-Dichloropyridine-4-methanol
Formula: C₆H₅Cl₂NO
Melting Pt: 127…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00052638
CAS Number: 101990-69-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR