(S)-(+)-2-(Methoxymethyl)pyrrolidine

Supplier: APOLLO SCIENTIFIC
Warning

OR4615-1G OR4615-250MG
76611-892EA 35.66 USD
76611-892 76611-894
(S)-(+)-2-(Methoxymethyl)pyrrolidine
(S)-(+)-2-(Methoxymethyl)pyrrolidine
Formula: C₆H₁₃NO
Boiling Pt: 61…62 °C/40mmHg
Density: 0.932 g/cm³ (25 °C)
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00010408
CAS Number: 63126-47-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR