2-Bromo-3-chloroaniline 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR51860-1G OR51860-25G OR51860-5G
76616-912EA 20.87 USD
76616-912 76616-914 76616-916
2-Bromo-3-chloroaniline 98%
2-Bromo-3-chloroaniline
Formula: C₆H₅BrClN
Melting Pt: 41…43 °C
Storage Temperature: Ambient
MDL Number: MFCD10699543
CAS Number: 96558-73-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR