Sulfur trioxide pyridine complex

Supplier: APOLLO SCIENTIFIC
Danger

OR24145-100G OR24145-500G
76620-842EA 38.72 USD
76620-842 76620-844
Sulfur trioxide pyridine complex
Sulphur trioxide pyridine complex
Formula: C₅H₅NO₃S
Melting Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00012437
CAS Number: 26412-87-3
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR