9-Fluorenylmethyl chloroformate (Fmoc chloride)
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR13210-100G
OR13210-25G
76615-630EA
34.64
USD
76615-630
76615-632
9-Fluorenylmethyl chloroformate (Fmoc chloride)
9-Fluorenylmethyl chloroformate (Fmoc chloride)
Formula:
C₁₅H₁₁ClO₂ Melting Pt: 62…64 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001138 CAS Number: 28920-43-6 EINECS: 249-313-6 UN: 2923 ADR: 8+6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...