Pyridinium tribromide 90+%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Pyridinium bromide perbromide, Monopyridin-1-ium tribromide, Pyridine hydrobromide perbromide

OR15023-100G OR15023-1KG OR15023-500G
76615-764EA 20.38 USD
76615-764 76615-766 76615-768
Pyridinium tribromide 90+%
Pyridinium tribromide
Formula: C₅H₆Br₃N
MW: 319.82 g/mol
Melting Pt: 127…133 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013223
CAS Number: 39416-48-3
EINECS: 254-446-8
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR