4-Ethynyltoluene

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Ethynyl-4-methylbenzene, p-Tolylacetylene

OR0642-25G OR0642-5G
76624-646EA 88.62 USD
76624-646 76624-648
4-Ethynyltoluene
4-Ethynyltoluene
Formula: C₉H₈
MW: 116.16 g/mol
Boiling Pt: 168…170 °C
Melting Pt: 23 °C
Density: 0.916 g/cm³ (25 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00008571
CAS Number: 766-97-2
UN: 3295
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR