2-(Aminomethyl)pyridine 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR5031-100G OR5031-25G
76611-968EA 41.76 USD
76611-968 76611-970
2-(Aminomethyl)pyridine 98%
2-(Aminomethyl)pyridine
Formula: C₆H₈N₂
Boiling Pt: 202…204 °C
Melting Pt: –20 °C
Density: 1.049 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00006360
CAS Number: 3731-51-9
EINECS: 223-090-5
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR