Phthalaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde

OR18349-100G OR18349-250G OR18349-25G
76615-904EA 77.42 USD
76615-904 76615-906 76615-908
Phthalaldehyde 98%
Phthalaldehyde
Formula: C₈H₆O₂
Boiling Pt: 83 °C/0.8mmHg
Melting Pt: 53…57 °C
Density: 1.13 g/cm³ (25 °C)
Flash Pt: 132 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003335
CAS Number: 643-79-8
EINECS: 211-402-2
UN: 2923
ADR: 8+6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR