tert-Butyl carbamate 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Carbamic acid tert-butyl ester
OR55005-25G
OR55005-500G
OR55005-100G
76592-176EA
20.87
USD
76592-176
76592-180
76592-178
tert-Butyl carbamate 98%
tert-Butyl carbamate
Formula:
C₅H₁₁NO₂ Melting Pt: 105…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007962 CAS Number: 4248-19-5 EINECS: 224-209-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...