1,10-Phenanthroline monohydrate

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1,10-Phenanthroline hydrate

OR2359-100G OR2359-250G OR2359-25G
76616-108EA 55.02 USD
76616-108 76616-110 76616-112
1,10-Phenanthroline monohydrate
1,10-Phenanthroline monohydrate
Formula: C₁₂H₈N₂·1H₂O
MW: 198.22 g/mol
Melting Pt: 97…104 °C
Density: 1.259 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149973
CAS Number: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR