Ruthenium(III) chloride hydrate 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Ruthenium(III) chloride xhydrate, Ruthenium trichloride hydrate

IN3062-1G IN3062-25G IN3062-5G
76615-098EA 38.46 USD
76615-098 76615-100 76615-102
Ruthenium(III) chloride hydrate 97%
Ruthenium (III) chloride hydrate
Formula: Cl₃Ru·xH₂O
MW: 225.44
Melting Pt: 100 °C (dec.)
Density: 3.11
Storage Temperature: Ambient
MDL Number: MFCD00149844
CAS Number: 14898-67-0
EINECS: 233-167-5
UN: 3260
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR